Jump to:
Product Details & Documents
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
- Fast analysis with reliable results
- Barcode calibration
- Low analysis costs
- Environmentally friendly
Documents
msds_116124
SDS
Specifications
Specifications
- Chemical method:Phthalein complexone
- Pk:50 Tests
- Description:Magnesium test
- Range:5 - 100 mg/l Mg
- Cat. No.:1.16124.0001
- M-Clarity quality level:MQ100
Specifications
Complementary Products
Product Information
- Catalog #DescriptionModelAvailabilityPrice
- 1.17246.0001Reflectometer for test strips onlyRQflex 20Log in for availability1 * 1 ST - SFr. 2’250.00