ContactUsLinkComponent
Test strips, glucose, Reflectoquant®, Supelco®
Test strips, glucose, Reflectoquant®, Supelco®
Catalog # 1.16720.0001
Supplier:  Merck
CAS Number:  
Test strips, glucose, Reflectoquant®, Supelco®
Catalog # 1.16720.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Chemical method:
    Enzymatic reaction
  • Pk:
    50 Tests
  • Description:
    Glucose test
  • M-Clarity quality level:
    MQ100
  • Range:
    1,0 - 100 mg/l C₆H₁₂O₆
Specifications
About this item
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly