ContactUsLinkComponent
Test strips, iron, Reflectoquant®, Supelco®
Katalognr. 1.16982.0001
Anbieter:  Merck
CAS-Nummer:  
Test strips, iron, Reflectoquant®, Supelco®
Katalognr. 1.16982.0001
Anbieter:  Merck
CAS-Nummer:  
Technische Daten
  • Chemical method:
    Triazine
  • Pk:
    50 Tests
  • Description:
    Iron test
  • M-Clarity quality level:
    MQ100
  • Range:
    0,5 - 20,0 mg/l Fe(II)
Technische Daten
Über diesen Artikel
Test strips for the reflectometers RQflex® 10, RQflex® 10 plus and RQflex® 20.
  • Fast analysis with reliable results
  • Barcode calibration
  • Low analysis costs
  • Environmentally friendly