Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Test kit, pH cell test, Spectroquant®, Supelco®

Test kit, pH cell test, Spectroquant®, Supelco®

pH cell test, pH 6,4 - 8,8, Indicator, Prove, Nova 30/60, Multy, Move, Picco, 280 Tests
Catalog # 1.01744.0001
Supplier:   Merck
Test kit, pH cell test, Spectroquant®, Supelco®
Catalog # 1.01744.0001
Supplier:  Merck
CAS Number:  
Test kit, pH cell test, Spectroquant®, Supelco®
Test kit, pH cell test, Spectroquant®, Supelco®

pH cell test, pH 6,4 - 8,8, Indicator, Prove, Nova 30/60, Multy, Move, Picco, 280 Tests

CAS Number:  
Price
SFr. 114.00
Log in for availability
1 * 280 Tests
Product Details & Documents

All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).

  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times

Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.

Specifications

Specifications

  • Chemical method:
    Indicator
  • Pk:
    280 Tests
  • Description:
    pH cell test
  • For unit:
    Prove, Nova 30/60, Multy, Move, Picco
  • Range:
    pH 6,4 - 8,8
  • Cat. No.:
    1.01744.0001
  • M-Clarity quality level:
    MQ100

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.