Order Entry
Switzerland
ContactUsLinkComponent
 
Visual colour kits, pH indicator solution with colour Card, pH 9,0 - 13,0, Supelco®
Catalog # 1.09176.0100
Supplier:  Merck
CAS Number:  
Visual colour kits, pH indicator solution with colour Card, pH 9,0 - 13,0, Supelco®
Catalog # 1.09176.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Description:
    pH indicator liquid
  • Seq:
    02
  • M-Clarity quality level:
    MQ200
  • Range:
    pH 9-10-11-12-13

Specifications

About this item

Quick tests, visual colour comparision.

  • Results in approximately 1 minute
  • Easy to use
  • Application-oriented measuring ranges
  • Portable

Titrimetric and colorimetric methods.

This range comprises both titrimetric and colorimetric tests for medium concentrations. With its broad range of available test parameters, these quick tests are the method of choice for aquaria, fish farming, aquatic ecology, environmental, cooling and boiler water analysis.

Delivery: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart and extensive instructions for use.