General purpose gloves, Soflex
About this item
Nitrile glove on interlock cotton base with ventilated back and knitted wrist. The nitrile coating ensures good chemical properties and outstanding mechanical properties (abrasion).
- Nitrile formula gives greater elasticity and unrivalled dexterity
- Cotton interlock lining strengthens the glove's mechanical performance while absorbing hand perspiration
- Ventilated back improves the glove's breathability
- Ribbed at the cuff, good wrist support is provided, thus eliminating risks of getting caught
Particularly suitable for the light and medium handling of oils, greases and hydrocarbon by-products.
Certifications: EN 388:2016 (3111X), Cat. II
3 Options Available Below
Product Details & Documents
Nitrile glove on interlock cotton base with ventilated back and knitted wrist. The nitrile coating ensures good chemical properties and outstanding mechanical properties (abrasion).
- Nitrile formula gives greater elasticity and unrivalled dexterity
- Cotton interlock lining strengthens the glove's mechanical performance while absorbing hand perspiration
- Ventilated back improves the glove's breathability
- Ribbed at the cuff, good wrist support is provided, thus eliminating risks of getting caught
Particularly suitable for the light and medium handling of oils, greases and hydrocarbon by-products.
Certifications: EN 388:2016 (3111X), Cat. II
Specifications for Product Family
Specifications
- Catalog No:
- 112-1514
- 112-1515
- 112-1511
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Finish:
- ¾ coating
- ¾ coating
- ¾ coating
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (3111X)
- EN 388:2016 (3111X)
- EN 388:2016 (3111X)
Specifications
Product Family Options
Product Information
- Glove sizeColourLengthThicknessAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.112-1514Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLiner materialCottonFinish¾ coatingCuff styleKnit wristEN normEN 388:2016 (3111X) - Specifications:
More
Specifications
Cat. No.112-1515Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLiner materialCottonFinish¾ coatingCuff styleKnit wristEN normEN 388:2016 (3111X) - Specifications:
More
Specifications
Cat. No.112-1511Ambidextrous / hand-specificHand-specificDisposable / reusableReusableLiner materialCottonFinish¾ coatingCuff styleKnit wristEN normEN 388:2016 (3111X)