ContactUsLinkComponent
Burette and thermometer clamp with shaft, AUTOMATIC
Burette and thermometer clamp with shaft, AUTOMATIC
Katalognr. BOCH5624
Supplier:  BOCHEM
undefined
Burette and thermometer clamp with shaft, AUTOMATIC
Katalognr. BOCH5624
Supplier:  BOCHEM
Specifications
  • For:
    Burettes and thermometers
  • Spanning width:
    0 - 25 mm
  • Pk:
    1
Specifications
About this item
This chromium plated clamp with shaft (length 115 mm) is used for holding burettes or thermometer (Ø 0 - 25 mm).