


Sulphuryl chloride 1.0 M in dichloromethane, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7791-25-5
:
Formula:Cl₂O₂S
MDL Number:MFCD00011451
:134.97 g/mol
Storage Temperature:Ambient
EINECS:232-245-6
UN:2922
ADR:8,II
:GHS05
:GHS06
:GHS08
Specification Test Results
- Appearance:Clear colorless to yellow Solution
- Titration Argentometric:0.95 to 1.10 M
Specification Test Results
Related Information
Frequently Bought Together