Organic reference standard, 4-Aminoazobenzene 100 µg/ml in acetonitrile, Dr Ehrenstorfer
Catalog # EHERXA10167500AL
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:60-09-3
- Description:4-Aminoazobenzene 100 µg/ml in acetonitrile
- Environmentally Preferable:
- Formula:C₁₂H₁₁N₃
- Description:4-Aminoazobenzene