Silver nitrate 2.395 g/l 0.0141 mol/l (0.0141 N) in aqueous solution
Catalog # HACH31653
Supplier: Hach
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:169.87 g/mol
Storage Temperature:Ambient
EINECS:231-853-9
CAS Number:7761-88-8
Formula:AgNO₃
MDL Number:MFCD00003414