Butyl phosphate (mixture of mono and diesters) for synthesis, Sigma-Aldrich®
: Merck
:12788-93-1
:
:8,III
:M-Clarity™ quality level = MQ200
:235-826-2
:1718
:GHS05
:Ambient
- Solubility:(20 °C) partly soluble
- Boiling Point:145 °C (1013 hPa)
- Flash Point:108.5 °C
- Density:1.13 g/cm3 (20 °C)
- Synonyms:Dibutyl phosphate, Phosphoric acid butyl ester
Frequently Bought Together