

2,4-Lutidine, TECHNICAL
25044.261
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Refractive index (20°C):1.498 - 1.500
:
:107.16 g/mol
:159 °C (1013 hPa)
:–60 °C
:0.93 g/cm³ (20 °C)
:50 °C
:Ambient
:203-586-8
:1992
:3 (6.1),III
:GHS02
:GHS06
:108-47-4
:C₇H₉N
:MFCD00006337