Tetrahydrofuran, Supelco®
: Merck
:109-99-9
:
:C₄H₈O
:MFCD00005356
:72.11 g/mol
:12,09351
:66 °C (1013 hPa)
:–108.5 °C
:0.888 g/cm³ (20 °C)
:Raumtemperatur
:203-726-8
:2056
:3,II
:M-Clarity™ quality level = MQ100
:–21.5 °C
:05-2114615188-48
:GHS02
:GHS08