Order Entry
Switzerland
ContactUsLinkComponent
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Flammable
Health
Irritant
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
undefined
Flammable
Health
Irritant
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier:  VWR Chemicals
Certificates
Limited Time Sale
Select items below are currently on sale

Specification Test Results

  • Assay:
    Min. 99 %
  • Appearance:
    Clear colourless liquid
  • IR Spectrum:
    Passes test
  • Boiling point:
    65.0 - 66.5 °C
  • Density (20/4):
    0.885 - 0.888
  • Free acidity:
    Max. 0.0003 meq/g
  • Refractive index (20°C):
    1.406 - 1.408
  • Peroxides (as H2O2):
    Max. 100 ppm
  • Water:
    Max. 0.03 %

Specification Test Results

Frequently Bought Together