General purpose gloves, Perfect Poly® Finger
Supplier: HONEYWELL SAFETY
Certificates
About this item
White polyamide knitted glove with white polyurethane coating on fingertips and an elastic wrist.
- Tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- Ventilated back and palm optimise the glove's breathability
- PU coating ensures a good grip at the fingertips
- Non pilling: Polyamide is suitable for precision work
Suitable for handling fine products in a dry environment. It is certified as 'silicone-free' to ensure its use in painting environments.
Certifications: Cat. I
Specifications
- Catalog No:
- DALL2232240/7
- DALL2232240/10
- DALL2232240/6
- DALL2232240/9
- DALL2232240/8
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Polyamide
- Finish:
- Coating on fingertips
- Coating on fingertips
- Coating on fingertips
- Coating on fingertips
- Coating on fingertips
- Gauge:
- 15
- 15
- 15
- 15
- 15
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist