ContactUsLinkComponent
Petri dish, Analyslide®
Petri dish, Analyslide®
Catalog # 516-8020
undefined
Petri dish, Analyslide®
Catalog # 516-8020
Specifications
  • Sterility:
    Non sterile
  • Version:
    Analyslide®
  • Pk:
    100
Specifications
About this item
The Analyslide® Petri dish has a rectangular base and a circular chamber that accepts a 47 mm membrane filter. The sample is protected during microscopic examination, and membrane filters can be stored and protected for reference.
  • Rectangular base fits most microscope stages
  • Inner cover ring for secure closure
  • Clear lid does not interfere with viewing sample
  • Frosted area on base permits identification of the sample with marker
  • Convenient box simplifies collection and storage of samples
Base and lid are constructed from PS.