Chromium(III) oxide, TECHNICAL
Plastic bottle for solids, 1 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Synonyms: Chromic oxide
- CAS Number:1308-38-9
- Formula:Cr₂O₃
- MDL Number:MFCD00010949
- Molecular Weight:151.99 g/mol
- Merck Index:13,02247
- Boiling Pt:4000 °C (1013 hPa)
- Melting Pt:2435 °C
- Density:5.22 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:215-160-9
Documents
Certificates
Search by Lot Number1L Plastic bottle for solids DIN 60 UN (ZPACK13251000)
Packaging Information
P20_VWRC22796.292_SPEC_EN.pdf spec_VWRC22796.292 spec_VWRC22796.292 spec_VWRC22796.292
Product specification
Specifications
Specification Test Results
- Identification:Passes test
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



