Silver nitrate, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7761-88-8
:
Formula:AgNO₃
MDL Number:MFCD00003414
:169.87 g/mol
Merck Index:12,08661
Boiling Pt:444 °C (1013 hPa)
Melting Pt:210 °C
Density:4.35 g/cm³ (20 °C)
UN:1493
ADR:5.1,II
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:231-853-9
REACH:01-2119513705-43
:GHS03
:GHS05
:GHS09
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Solubility:2160 g/l (20 °C)
- Melting Point:212 °C
- Molar Mass:169.87 g/mol
- Bulk Density:2350 kg/m3
- Boiling Point:444 °C (1013 hPa) (decomposition)
- Density:4.35 g/cm3 (20 °C)
- pH:5.4 - 6.4 (100 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Specification Test Results
Frequently Bought Together