


Selenium disulphide
Catalog # MOLEM36260646
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:143.09 g/mol
Boiling Pt:–112 °C (1013 hPa)
Density:0.03 g/cm³ (–65 °C)
Storage Temperature:Ambient
EINECS:231-303-8
UN:2657
ADR:6.1,II
:GHS06
:GHS08
:GHS09
CAS Number:7488-56-4
Formula:S₂Se
MDL Number:MFCD00011227