Diphosphorus pentoxide ACS, ISO, Reag. Ph. Eur. analytical reagent, Supelco®
: Merck
:1314-56-3
:
:P₂O₅
:MFCD00011440
:141.94 g/mol
:13,07441
:562 °C
:2.253 g/cm³ (20 °C)
:215-236-1
:8,II
:M-Clarity™ quality level = MQ200
:1807
:GHS05
:Ambient
Frequently Bought Together