


Carbon disulphide 99.9%, extra pure low in benzene
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:75-15-0
:
Formula:CS₂
MDL Number:MFCD00011321
:76.14 g/mol
Merck Index:13,01821
Boiling Pt:46.2 °C (1013 hPa)
Melting Pt:–111 °C
Density:1.262 g/cm³
Flash Pt:–30 °C
Storage Temperature:Ambient
EINECS:200-843-6
UN:1131
ADR:3,I
:GHS07
:GHS08
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Assay:≥99.85 % (By difference)
- Residue after evaporation:≤20 ppm
- Hydrogen sulfide (H2S):≤3 ppm
- Benzene:≤50 ppm
- HPLC:≥97.5 %
Specification Test Results
Frequently Bought Together