Order Entry
Switzerland
ContactUsLinkComponent
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh), extra pure for column chromatography, Millipore®
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh), extra pure for column chromatography, Millipore®
Catalog # 1.07754.0500
CAS Number:  
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh), extra pure for column chromatography, Millipore®
Catalog # 1.07754.0500
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • pH-value (10 % suspension):
    6.0 - 7.5
  • Chloride (Cl):
    ≤ 0.008 %
  • Sulphate (SO₄):
    ≤ 0.003 %
  • Cd (Cadmium):
    ≤ 0.0005 %
  • Cu (Copper):
    ≤ 0.0005 %
  • Fe (Iron):
    ≤ 0.002 %
  • Pb (Lead):
    ≤ 0.0005 %
  • Zn (Zinc):
    ≤ 0.0005 %
  • Flow time:
    60 s / 10 cm
  • Particle size:
    90% between 0.063-0.200 mm
  • Loss on drying (150 °C):
    ≤ 7.0 %
  • UV absorption (Dichloro methane extract):
    conforms

Specification Test Results

About this item

 :
 :60.08 g/mol
Storage Temperature:Ambient
EINECS:231-545-4
CAS Number:63231-67-4
 :7631-86-9
Formula:SiO₂
MDL Number:MFCD00011232