Order Entry
Switzerland
ContactUsLinkComponent
Silica gel, Pore size: 60 Å <0.063 mm for column chromatography, Millipore®
Silica gel, Pore size: 60 Å <0.063 mm for column chromatography, Millipore®
Catalog # 1.07729.1000
CAS Number:  
Silica gel, Pore size: 60 Å <0.063 mm for column chromatography, Millipore®
Catalog # 1.07729.1000
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • pH-value (10 % suspension):
    6.5 - 7.5
  • Fe (Iron):
    ≤ 0.02 %
  • Loss on drying:
    ≤ 7.0 %
  • Particle size:
    95 %  0.063 mm

Specification Test Results

About this item

 :
 :60.08 g/mol
Storage Temperature:Ambient
EINECS:231-545-4
CAS Number:63231-67-4
 :112926-00-8
Formula:SiO₂
MDL Number:MFCD00011232