



Tin(II) chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7772-99-8
:
Formula:SnCl₂
MDL Number:MFCD00011241
:189.62 g/mol
Merck Index:13,08861
Boiling Pt:623 °C (1013 hPa)
Melting Pt:246 °C
Density:3.95 g/cm³ (20 °C)
UN:3260
ADR:8,II
:M-Clarity™ quality level = MQ200
EINECS:231-868-0
:GHS05
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Solubility:2700 g/l (20 °C)
- Melting Point:246 °C
- Molar Mass:189.62 g/mol
- Boiling Point:623 °C (1013 hPa)
- Density:3.95 g/cm3 (20 °C)
- pH:2.0 (100 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Frequently Bought Together