Order Entry
Switzerland
ContactUsLinkComponent
Diboron trioxide for analysis of silicates, Supelco®
Health
Diboron trioxide for analysis of silicates, Supelco®
Catalog # 1.00163.1000
Supplier:  Merck
CAS Number:  
Health
Diboron trioxide for analysis of silicates, Supelco®
Catalog # 1.00163.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Solubility:
    36 g/l (25 °C)
  • Melting Point:
    450 °C
  • Molar Mass:
    69.61 g/mol
  • Bulk Density:
    900 - 1100 kg/m3
  • Boiling Point:
    1860 °C
  • Density:
    2.46 g/cm3 (20 °C)
  • pH:
    4 (10 g/l, H2O, 25 °C)

Specification Test Results

About this item

 :
 :69.62 g/mol
Merck Index:13,01327
Boiling Pt:1860 °C (1013 hPa)
Melting Pt:450 °C
Density:1.81…1.86 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:215-125-8
REACH:01-2119486655-24
 :GHS08
CAS Number:1303-86-2
Formula:B₂O₃
MDL Number:MFCD00011315