Thallium(I) chloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Thallium chloride
CAS Number:7791-12-0
:
Formula:ClTl
MDL Number:MFCD00011274
Boiling Pt:720
Melting Pt:430
Density:7 g/cm³
Molecular Weight:239.84 g/mol
EINECS:232-241-4
ADR:6.1 II
Merck Index:12,09395
UN:1707
:GHS06
:GHS08
:GHS09
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to light yellow Powder
- Titration Argentometric:98.5 to 101.5 %
Specification Test Results
Frequently Bought Together