
Ammonium fluoride 40% in aqueous solution
Catalog # 40292-5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Plastic can
- Pk:5 L
- Seq:5190
Specifications
About this item
:
:37.04 g/mol
Storage Temperature:Ambient
EINECS:235-185-9
UN:3287
ADR:6.1,III
:GHS06
CAS Number:12125-01-8
Formula:NH₄F
MDL Number:MFCD00011423