
Anthrone in ACS Reagent grade water 97%, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:90-44-8
:
Formula:C₁₄H₁₀O
MDL Number:MFCD00001187
Melting Pt:154...157 °C (lit.)
:M-Clarity™ quality level = MQ200
:194.23 g/mol
EINECS:201-994-0
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (color):Off white to yellow
- Appearance (form):Powder or crystals
- Melting point:5 degrees celsius range (maximum), including 156 degrees c
- Solubility (color):Faint yellow to dark yellow and faint green-yellow to dark green-yellow
- Solubility (turbidity):Clear
- Solubility (method):2% in ethyl acetate
- Carbon content:83.5 - 89.6 %
- Infrared spectrum:Conforms to structure
- Wavelength (1) (UV):425 nm, passes absorbance of reagent solution test
- Absorbance (1):≤ 0.75
- Wavelength (2) (UV):620 nm, passes absorbance of reagent solution test
- Absorbance (2):≤ 0.045
- Solvent (UV):Sulfuric acid (acc. To acs-test absorbance of reagent solution)
- Miscellaneous assay:Passes test sensitivity to carbohydrates
- ACS specifications:Passes
Specification Test Results
Frequently Bought Together