

Ammonia solution 5.0 M NH4OH (5.0N) volumetric solution, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Boiling Pt:38 °C
Density:0.9…1.0 g/cm³
Storage Temperature:Ambient
EINECS:215-647-6
ADR:8,III
:GHS07
:GHS05
Specification Test Results
- Assay:4.90 - 5.10 N
- Traceable to NIST SRM:effective
- Explosion Limit:27 %
Specification Test Results
Frequently Bought Together