

N,N-Dimethylallylamin, Sigma-Aldrich®
: Merck
:2155-94-4
:
:C₅H₁₁N
:MFCD00038234
:85.15 g/mol
:3 (8),II
:M-Clarity™ quality level = MQ200
:218-458-7
:2733
:GHS02
:GHS07
:Raumtemperatur
- Solubility:(20 °C) soluble
- Molar Mass:85.14 g/mol
- Boiling Point:62 - 64 °C
- Flash Point:8 °C
- Refractive Index:1.3998 (20 °C, 589 nm)
- Density:0.730 g/cm3 (20 °C)
Frequently Bought Together