Order Entry
Switzerland
ContactUsLinkComponent
2-Hydroxypropyl methacrylate (mixture of isomers) ≥97% stabilised, Sigma-Aldrich®
Irritant
2-Hydroxypropyl methacrylate (mixture of isomers) ≥97% stabilised, Sigma-Aldrich®
Supplier:  Merck
Certificates
Irritant
2-Hydroxypropyl methacrylate (mixture of isomers) ≥97% stabilised, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Assay isomers (GC, area%):
    ≥ 97.0 % (a/a)
  • Density (d 20 °C/ 4 °C):
    1.027 - 1.030
  • Identity (IR):
    passes test

Specification Test Results

Related Information

Frequently Bought Together