


Ammonia solution 28.0 - 30.0% (NH₃ basis), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:1336-21-6
:7664-41-7
Formula:NH₃
MDL Number:MFCD00066650
:35.05 g/mol
Boiling Pt:32…37.7 °C (1013 hPa)
Melting Pt:–60…–57.5 °C
Density:0.88…0.91 g/cm³ (20 °C)
EINECS:215-647-6
UN:2672
ADR:8,III
:GHS05
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Meets ACS Reagent Chemical Requirements:—
- For Laboratory,Research,or Manufacturing Use:—
- Appearance (Colorless and free from suspended matter or sediment):Passes Test
- Assay (as NH₃):28.0 - 30.0 %
- Color (APHA):≤5
- Specific Gravity at 60°/60°F:0.896 - 0.902
- Residue after Ignition:≤0.0020 %
- Carbon Dioxide (CO₂):≤0.002 %
- Substances Reducing Permanganate:Passes Test
- Chloride (Cl):≤0.5 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤2 ppm
- Sulfate (SO₄):≤2 ppm
- Trace Impurities - Aluminum (Al):≤200.0 ppb
- Arsenic and Antimony (as As):≤3000.0 ppb
- Trace Impurities - Barium (Ba):≤300.0 ppb
- Trace Impurities - Boron (B):≤50 ppb
- Trace Impurities - Chromium (Cr):≤100.0 ppb
- Trace Impurities - Copper (Cu):≤100.0 ppb
- Trace Impurities - Gold (Au):≤200.0 ppb
- Heavy Metals (as Pb):≤500.0 ppb
- Trace Impurities - Iron (Fe):≤100.0 ppb
- Trace Impurities - Lead (Pb):≤200.0 ppb
- Trace Impurities - Magnesium (Mg):≤200.0 ppb
- Trace Impurities - Manganese (Mn):≤100.0 ppb
- Trace Impurities - Nickel (Ni):≤100.0 ppb
- Trace Impurities - Tin (Sn):≤100 ppb
- Trace Impurities - Titanium (Ti):≤100.0 ppb
- Trace Impurities - Zinc (Zn):≤100.0 ppb
- Meets Reagent Specifications for testing USP/NF monographs:—
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_BAKR9721-03 spec_BAKR9721-06 spec_BAKR9721-15
Product specification
Frequently Bought Together