Orders LinkContactUsLinkComponent
Slide folder
Slide folder
Preparation folders, Natural, Cardboard, 100×80×6 mm, 2
Catalog # MARI6001000
Supplier:   MARIENFELD
Slide folder
Catalog # MARI6001000
Supplier:  MARIENFELD
Slide folder
Slide folder
Preparation folders, Natural, Cardboard, 100×80×6 mm, 2
Price
SFr. 211.00
Log in for availability
1 * 50 ST
Product Details & Documents
Preparation pasteboards with cover and recessed grips.
Specifications
  • No. of slides:
    2
  • Colour:
    Natural
  • Description:
    Preparation folders
  • Material:
    Cardboard
  • W×D×H:
    100×80×6 mm