Orders LinkContactUsLinkComponent
Test tubes with rim
Test tubes with rim
Supplier:   MARIENFELD
Test tubes with rim
Supplier:  MARIENFELD
Certificates
Test tubes with rim
Test tubes with rim
2 Options Available Below
Product Details & Documents
Soda-Lime glass.
  • Very good chemical resistance
  • Good temperature resistance
Product Family Options
Product Information
  • Capacity
    Length
    Ext.Ø
    Wall thickness
    Availability
    Price
  • 4 ml
    75 mm
    10 mm
    0,6 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - SFr. 111.00
    SFr. 111.00
    1 * 1.000 ST
       SFr. 111.00
  • 9 ml
    100 mm
    12 mm
    0,6 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - SFr. 60.30
    SFr. 60.30
    1 * 500 ST
       SFr. 60.30