Capillary columns, BPX35
Supplier: Trajan Scientific and Medical
Certificates
About this item
These columns are suitable for environmental analyses, pesticides/herbicides, drugs of abuse, pharmaceuticals, polynuclear aromatic hydrocarbons and GC-MS applications.
35% phenyl polysilphenylene-siloxane
Max. temperature: 10 to 330/360 °C
Suitable replacement for: DB-35, DB-35ms, Rtx-35, HP-35, HP-35MS, SPB-35, MDN-35.
Frequently Bought Together