
Sulphuric acid ≥98% for nitrogen analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7664-93-9
:
Formula:H₂SO₄
MDL Number:MFCD00064589
:98.08 g/mol
Merck Index:14,08974
Boiling Pt:∼335 °C (1013 hPa)
Melting Pt:3 °C
Density:1.84 g/cm³ (20 °C)
UN:1830
ADR:8,II
:M-Clarity™ quality level = MQ300
EINECS:231-639-5
:GHS05
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specification Test Results
- Assay (alkalimetric):≥ 97.5 %
- Total nitrogen (N):≤ 1 ppm
- Residue on ignition:≤ 2 ppm
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.00748.0500_TDS_EN ProductPage_100748
Product specification
msds_100748
SDS
Frequently Bought Together