
Sulphuric acid 95-98%, BAKER ANALYZED® ACS, J.T.Baker®
: Avantor
:7664-93-9
:
:H₂SO₄
:MFCD00064589
:98.08 g/mol
:14,08974
:∼335 °C (1013 hPa)
:3 °C
:1.84 g/cm³ (20 °C)
:Ambient
:231-639-5
:1830
:8,II
:GHS05
- Assay:95.0 - 98.0 %
- Ammonium (NH4):max. 1 ppm
- Appearance:passes test
- Color (APHA):max. 10
- Mercury (Hg):max. 5 ppb
- Residue after Ignition:max. 4 ppm
- Substances Reducing KMnO4(as SO2):max. 2 ppm
- Silver (Ag):max. 0.01
- Aluminium (Al):max. 0.05
- Arsenic (As):max. 0.01
- Barium (Ba):max. 0.02
- Beryllium (Be):max. 0.01
- Calcium (Ca):max. 0.5
- Cadmium (Cd):max. 0.02
- Chloride (Cl):max. 0.1
- Cobalt (Co):max. 0.01
- Chromium (Cr):max. 0.1
- Copper (Cu):max. 0.01
- Iron (Fe):max. 0.1
- Germanium (Ge):max. 0.05
- Potassium (K):max. 0.1
- Lithium (Li):max. 0.01
- Magnesium (Mg):max. 0.1
- Manganese (Mn):max. 0.01
- Molybdenum (Mo):max. 0.02
- Sodium (Na):max. 0.5
- Nickel (Ni):max. 0.05
- Lead (Pb):max. 0.05
- Strontium (Sr):max. 0.01
- Titanium (Ti):max. 0.1
- Thallium (Tl):max. 0.05
- Vanadium (V):max. 0.01
- Zinc (Zn):max. 0.05
- Zirconium (Zr):max. 0.1
- Heavy Metals (as Pb):max. 1
- Nitrate (NO3):max. 0.5
Frequently Bought Together