Order Entry
Switzerland
ContactUsLinkComponent
Sodium sulphite 95.0-100.5%, purified
Sodium sulphite 95.0-100.5%, purified
Supplier:  VWR Chemicals
Certificates
undefined
Sodium sulphite 95.0-100.5%, purified
Supplier:  VWR Chemicals
Certificates

Specification Test Results

  • Assay (at filling):
    95.0 - 100.5 %
  • Heavy metals (as Pb):
    Max. 50 ppm
  • Fe (Iron):
    Max. 20 ppm

Specification Test Results

Frequently Bought Together