


Sodium peroxodisulphate
Supplier: VWR Collection
Certificates
About this item
CAS Number:7775-27-1
:
Formula:Na₂S₂O₈
MDL Number:MFCD00003501
:238.11 g/mol
Merck Index:13,08729
Melting Pt:100 °C
Density:2.4 g/cm³ (20 °C)
EINECS:231-892-1
UN:1505
ADR:5.1,III
:GHS03
:GHS08
:GHS07
Storage Temperature:Ambient
Frequently Bought Together