


Sodium peroxodisulphate, EMSURE® for analysis, Supelco®
Supplier: Merck
Certificates
About this item
CAS Number:7775-27-1
:
Formula:Na₂S₂O₈
MDL Number:MFCD00003501
:238.11 g/mol
Merck Index:13,08729
Melting Pt:100 °C
Density:2.4 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-892-1
ADR:5.1,III
:M-Clarity™ quality level = MQ300
UN:1505
:GHS03
:GHS08
:GHS07
Specification Test Results
- Assay (iodometric):≥ 99.0 %
- Identity:passes test
- Chloride (Cl):≤ 0.01 %
- Heavy metals (as Pb):≤ 0.003 %
- NH₄ (Ammonium):≤ 0.05 %
- Cr (Chromium):≤ 0.001 %
- Cu (Copper):≤ 0.001 %
- Fe (Iron):≤ 0.0005 %
- Mn (Manganese):≤ 0.0001 %
- Pb (Lead):≤ 0.001 %
- Loss on drying (105 °C; 3 hours):≤ 0.05 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.06609.0500_TDS_EN MERC1.06609.1000_TDS_EN MERC1.06609.9025_TDS_EN ProductPage_106609
Product specification
msds_106609
SDS
Frequently Bought Together