Order Entry
Switzerland
ContactUsLinkComponent
 
Sodium metabisulphite ≥97.0-100.0% (by iodometry), EMPROVE® ESSENTIAL NF, JP, Ph. Eur., BP, E223, extra pure suitable for use as excipient, SAFC®
 
undefined
Sodium metabisulphite ≥97.0-100.0% (by iodometry), EMPROVE® ESSENTIAL NF, JP, Ph. Eur., BP, E223, extra pure suitable for use as excipient, SAFC®
 

 

  • Environmentally Preferable:

 

 

  • Assay (iodometric):
    97.0 - 100.0 %
  • Assay (iodometric, SO₂):
    65.4 - 67.4 %
  • Identity:
    passes test
  • Appearance of solution:
    passes test
  • pH-value (5 %; water):
    3.5 - 5.0
  • Chloride (Cl):
    ≤ 0.01 %
  • Heavy metals (as Pb):
    ≤ 0.001 %
  • Thiosulfate (S₂O₃):
    ≤ 0.05 %
  • As (Arsenic):
    ≤ 0.0002 %
  • Fe (Iron):
    ≤ 0.0005 %
  • Hg (Mercury):
    ≤ 0.0001 %
  • Pb (Lead):
    ≤ 0.0002 %
  • Se (Selenium):
    ≤ 0.0003 %
  • Residual solvents (Ph Eur/USP/ICH):
    excluded by the manufacturing process

 

Frequently Bought Together