Order Entry
Switzerland
ContactUsLinkComponent
Sodium metaphosphate, TECHNICAL
Sodium metaphosphate, TECHNICAL
Catalog # 27880.296
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium metaphosphate, TECHNICAL
Catalog # 27880.296
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • di-Phosphorus pentoxide:
    Min. 60 %

Specification Test Results

About this item

 :
 :101.96 g/mol
Melting Pt:625 °C
Density:2.54 g/cm³ (20 °C)
EINECS:233-782-9
CAS Number:10361-03-2
Formula:NaPO₃