Pasteur pipettes, disposable
Supplier: MARIENFELD
Jump to:
About this item
Pasteur pipettes made of soda-lime glass.
- For single use
- Long, fine tip
- Constricted upper suction end to retain a cotton plug (supplied without cotton plug)
- Handy packaging in boxes containing 250 pieces
: In compliance with DIN ISO 7712
2 Options Available Below
Product Details & Documents
Pasteur pipettes made of soda-lime glass.
- For single use
- Long, fine tip
- Constricted upper suction end to retain a cotton plug (supplied without cotton plug)
- Handy packaging in boxes containing 250 pieces
: In compliance with DIN ISO 7712
Product Family Options
Product Information
- Ext.ØLengthPackedAvailabilityPrice
- MARI3233049Pasteur pipette, unpluggedPasteur pipette, unplugged1,3 - 1,4 mm150 mm4×250 piecesLog in for availability1 * 1.000 STSFr. 143.00
- MARI3233050Pasteur pipette, unpluggedPasteur pipette, unplugged1,3 - 1,4 mm230 mm4×250 piecesLog in for availability1 * 1.000 STSFr. 122.00