

Sodium peroxide, granulated ACS, ISO analytical reagent, Supelco®
: Merck
:1313-60-6
:
:Na₂O₂
:MFCD00003497
:77.98 g/mol
:13,08728
:660 °C
:2.805 g/cm³ (20 °C)
:Ambient
:215-209-4
:5.1,I
:M-Clarity™ quality level = MQ100
:1504
:GHS03
:GHS05
- Solubility:100 g/l (20 °C) (decomposition)
- Melting Point:660 °C
- Molar Mass:77.98 g/mol
- Bulk Density:1138 kg/m3
- Density:2.8 g/cm3 (20 °C)
- pH:12.8 (100 g/l, H2O, 20 °C)
- Hazard Class:5.1
- Hazard Class Description:Oxidizer
Frequently Bought Together