ContactUsLinkComponent
Sodium peroxide ACS
Corrosive
Oxidizing
Sodium peroxide ACS
Certificates
undefined
Corrosive
Oxidizing
Sodium peroxide ACS
Specification Test Results
  • Appearance:
    Yellow Powder or granules (May contain a small quantity of discolored material from the manufacturing process)
  • Titration with KMnO4:
    ≥93.0 %
  • Chloride (Cl):
    ≤20 ppm
  • Phosphate (PO4):
    ≤5 ppm
  • Sulfate (SO4):
    ≤10 ppm
  • Iron (Fe):
    ≤50 ppm
  • Heavy metals (as Pb):
    ≤20 ppm