Screw caps for bottles with GL threads, BOLA
Supplier: Bohlender
Certificates
About this item
Screw cap with handy knurl, made of PTFE.
- Very good chemical resistance
- Temperature resistant from −200 to +250 °C
Suitable for bottles and vessels with GL threads.
Frequently Bought Together