Orders LinkContactUsLinkComponent
Water
Marketsource
 :   Roth Carl
Water
 :  Roth Carl
Marketsource
Water
 :  Hydrogen oxide
  •  :7732-18-5
  •  :H₂O
  •  :MFCD00011332
  •  :18.02 g/mol
  •  :13,10098
  •  :100 °C (1013 hPa)
  •  :0 °C
  •  :1 g/cm³ (4 °C)
  •  :Ambient
  •  :231-791-2
Product Family Options
  • Pack type
    Pk
  • Glass bottle
    1 L
    1 * 1 L
    1 * 1 L - SFr. 10.70
    SFr. 10.70
    1 * 1 L
       SFr. 10.70
    Marketsource
    Marketsource
  • 10 L
    1 * 10 L
    1 * 10 L - SFr. 46.70
    SFr. 46.70
    1 * 10 L
       SFr. 46.70
    Marketsource
    Marketsource
  • 30 L
    1 * 30 L
    1 * 30 L - SFr. 121.80
    SFr. 121.80
    1 * 30 L
       SFr. 121.80
    Marketsource
    Marketsource