Orders LinkContactUsLinkComponent
Hose connectors
Hose connectors
Supplier:   Bohlender
Hose connectors
Supplier:  Bohlender
Certificates
Hose connectors
5 Options Available Below
Product Details & Documents
These connectors are specially designed for a higher flow rate. Connectors made of PP, with elastic sealing lip, FPM o-ring and nut made of PBTP.
  • Available as straight or bent type
  • Universal chemical resistance
  • Maximum temperature up to 250 °C
  • Supplied with matching PBTP cap
Product Family Options
Straight
Product Information
  • Ext.Ø
    Ø int.
    Length
    Thread
    Availability
    Price
  • 10,8 mm
    7,0 mm
    52 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 6.10
    SFr. 6.10
    1 * 1 ST
       SFr. 6.10
  • 8,8 mm
    6,0 mm
    46 mm
    GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 10.10
    SFr. 10.10
    1 * 1 ST
       SFr. 10.10
  • 16,0 mm
    10,0 mm
    68 mm
    GL 25
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 19.30
    SFr. 19.30
    1 * 1 ST
       SFr. 19.30
Bent
Product Information
  • Ext.Ø
    Ø int.
    Length
    Thread
    Availability
    Price
  • 10,8 mm
    7,0 mm
    66 mm
    GL 18
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 5.10
    SFr. 5.10
    1 * 1 ST
       SFr. 5.10
  • 8,8 mm
    6,0 mm
    51 mm
    GL 14
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 11.10
    SFr. 11.10
    1 * 1 ST
       SFr. 11.10