Wasser, BAKER ANALYZED®, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:7732-18-5
:
Formula:H₂O
MDL Number:MFCD00011332
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
Storage Temperature:Raumtemperatur
EINECS:231-791-2
Specifications
- Environmentally Preferable:Unbedenklicher für Mensch und Umwelt
- Umweltzertifizierung:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
- Residue after Evaporation:max. 5 ppm
- at 210 nm:max. 0.005
- at 254 nm:max. 0.001
- Positive ESI-MS Sensitive Impurities (as Reserpine):max. 100 ng/ml
- Aluminium (Al):max. 500
- Calcium (Ca):max. 100
- Iron (Fe):max. 100
- Potassium (K):max. 100
- Magnesium (Mg):max. 100
- Sodium (Na):max. 100
Specification Test Results
Related Information
Certificates
Search by Lot Numbersds_BAKR9825
SDS
VWR J.T.Baker® High Purity solvents and reagents
Product Literature
Frequently Bought Together