Water, BAKER ANALYZED ULTRA LC/MS™, ULTRA for LC-MS, tested for UHPLC, J.T.Baker®
Catalog # 9823-02
Supplier: Avantor
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:EPA Safer Chemical Solvent (Green Circle Rating); Chem21 Solvent Selection Guide recommended by default and After discussion
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:Passes Test
- Appearance:Passes Test
- Gradient Elution Test (a.u.) - 254 nm:≤0.002
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.1 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤0.2 ppb
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine):≤25.0 ppb
- LC/MS Suitability - Largest Response on ESI- Negative Mode (as 4-Nitrophenol):≤25.0 ppb
- Trace Impurities - Aluminum (Al):≤10.0 ppb
- Trace Impurities - Calcium (Ca):≤20 ppb
- Trace Impurities - Chromium (Cr):≤5.0 ppb
- Trace Impurities - Cobalt (Co):≤5.0 ppb
- Trace Impurities - Copper (Cu):≤5.0 ppb
- Trace Impurities - Iron (Fe):≤5.0 ppb
- Trace Impurities - Lead (Pb):≤5.0 ppb
- Trace Impurities - Lithium (Li):≤5.0 ppb
- Trace Impurities - Magnesium (Mg):≤10.0 ppb
- Trace Impurities - Manganese (Mn):≤5.0 ppb
- Trace Impurities - Nickel (Ni):≤5.0 ppb
- Trace Impurities - Potassium (K):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤20.0 ppb
- Trace Impurities - Tin (Sn):≤5.0 ppb
- Trace Impurities - Zinc (Zn):≤5.0 ppb
Specification Test Results
About this item
Synonyms: Hydrogen oxide
:
:18.02 g/mol
Merck Index:13,10098
Boiling Pt:100 °C (1013 hPa)
Melting Pt:0 °C
Density:1 g/cm³ (4 °C)
:Filtered through a 0.1 micron filter.
Storage Temperature:Ambient
EINECS:231-791-2
CAS Number:7732-18-5
Formula:H₂O
MDL Number:MFCD00011332